Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
Product Code. 10509151 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Brown
Gray
Green
Green/Yellow
Red
White
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10509151 White Each
10410083 Green/Yellow Each
10459502 Green Each
10358082 Brown Each
10039500 Red Each
10568761 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10509151 Supplier Thermo Scientific™ Supplier No. 75008185
  • ONLINE EXCLUSIVE PRICE
    102.00€
    93.00€ / pezzi 1
    Risparmia 9.00€
    8% Sconto
In stock
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 4 x 180 Swing bucket rotor
Color White