missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG Series Glass Door Laboratory Refrigerators

Codice prodotto. 18938751 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Capacità (sistema imperiale):
12 cu. ft.
23 cu. ft.
29.2 cu. ft.
45.8 cu. ft.
51.1 cu. ft.
Interno:
White (Painted)
Requisiti elettrici:
208-230V/60Hz
230V/50Hz
Dimensione della confezione:
1 unità
Pezzo
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema imperiale) Interno Requisiti elettrici Dimensione della confezione
18938751 51.1 cu. ft. White (Painted) 208-230V/60Hz Pezzo
17876298 12 cu. ft. White (Painted) 230V/50Hz 1 unità
17866298 23 cu. ft. White (Painted) 230V/50Hz 1 unità
17896298 29.2 cu. ft. White (Painted) 230V/50Hz 1 unità
17886298 45.8 cu. ft. White (Painted) 230V/50Hz 1 unità
17846298 51.1 cu. ft. White (Painted) 230V/50Hz 1 unità
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18938751 Fornitore Thermo Scientific™ N. del fornitore TSG5005GD
  • ONLINE EXCLUSIVE PRICE
    10658.08€
    9321.05€ / pezzi 1
    Risparmia 1337.03€
    12% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 26-05-2027
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The TSG Glass Door Refrigerators offer sustainability at 2-8°C, an outstanding 4-year parts and labor warranty, and tight temperature uniformity in single and double door configurations. All TSG Series utilize SNAP and F-Gas compliant refrigeration and are made at a zero waste to landfill facility.

Thermo Scientific TSG Series Glass Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.

Thermo Scientific TSG Series Glass Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.

The drive for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • Stainless Steel interior finish available
  • E-Lock Adapter kits
TRUSTED_SUSTAINABILITY

Specifica

Descrizione TSG Series General Purpose Refrigerator
Tipo General Purpose Refrigerator
Capacità (sistema imperiale) 51.1 cu. ft.
Capacità (sistema metrico) 1447 L
Gamma di temperatura 3°C to 7°C
Temperatura (punto di regolazione) +5°C
Refrigerante R290
Door Style Glass
No. of Doors 2
Ripiani 8 Adjustable
Capacità (sistema imperiale) ripiano 100 lb.
Capacità (sistema metrico) ripiano 45 kg
Materiale armadietto Painted Steel
Finitura esterna Painted
Interno White (Painted)
Certificazione/Conformità cULus
Stile Upright
Tipo di controllo Capacitive Touch
Porte 1 Rear Access Port
Warranty 4 Years Parts and Labor + 1 Year Compressor
Opzioni di monitoraggio SmartVue Pro, Remote Alarm Contacts
Sicurezza dell'accesso Key Lock
Compressore Hermetic
Dimensioni esterno (PxLxA) 37.8 x 56.5 x 78.5 in. (960 x 1435 x 1994 mm)
Dimensioni interne (L x P x A) 28.5 x 52.5 x 58.0 in. (724 x 1334 x 1473 mm)
Tipo di spina NEMA 6-15
Tensione 208/230 V
Requisiti elettrici 208-230V/60Hz
Frequenza 60 Hz
Peso di spedizione (sistema imperiale) 669 lb.
Peso di spedizione (sistema metrico) 303 kg
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.