Learn More
Thermo Scientific™ ThermoChill II Recirculating Chillers
Codice prodotto. 15331657 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità di raffreddamento:
Up to 900 watts
Portata:
4.5 L/min.
7.6 L/min.
9.4 L/min.
Comunicazioni:
None
RS232
Tipo di pompa:
MDC1 (Mag Drive Centrifugal Pump)
PD1 (Positive Displacement Pump)
PD2 (Positive Displacement Pump)
Dimensione della confezione:
Pezzo
Codice prodotto. 15331657
Marca: Thermo Scientific™ 197222010000
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Choose these recirculating chillers for reliable liquid temperature control, with cooling capacity up to 1000W/3415 BTU.
Reliable liquid temperature control, with cooling capacity up to 1000W/3415 BTU. Thermo Scientific™ ThermoChill II Recirculating Chillers are a compact line of refrigerated recirculators ranging in cooling capacities from 600 to 2000 watts. Innovative design features such as minimized plumbing connections and an integrated full-flow fluid strainer maximize product reliability. With simplicity in design and operation, the ThermoChill platform will provide years of reliable liquid temperature control to meet all of your conventional application requirements.
  • Accessible fill port for easy filling
  • Pressure gauge located on control panel for easy viewing
  • Intuitive digital controller
  • Controller indicator lights illuminate when adjusting settings or when high/low temp alarms activated
  • Removable full-low strainer ensures clean recirculating fluid
  • Locking caster for easy unit maneuvering
  • RS232 or RS485 digital communications optional
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE marked/UL listed
Comunicazioni RS232
Capacità di raffreddamento Up to 900 watts
Portata 9.4 L/min.
Tipo di pompa PD2 (Positive Displacement Pump)
Modello ThermoChill II LR
Refrigerante R134A
Temperatura (punto di regolazione) -10°C to +30°C
Stabilità termica ±°
Electrical Requirements 230 V/50 Hz
Tensione 230 V
Tipo di collegamento RS 232
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for 1 year thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.