Learn More
Brannan™ Thermometer Science Kit
Codice prodotto. 13018573
Cambia vista
Click to view available options
Product Type:
Investigative Science Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
13018573 Investigative Science Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 13018573 Fornitore Brannan™ N. del fornitore 25/701/0
  • ONLINE EXCLUSIVE PRICE
    95.40€
    87.66€ / 1 sets
    Risparmia 7.74€
    8% Sconto
Spedizione stimata: 30-12-2026
Brannan™ Thermometer Science Kit comes in a blue plastic case and includes a variety of instruments and a set of investigative science notes.
Complete kit designed to assist teachers in fulfilling the national curriculum
Includes:
  • Blue plastic case (334 x 232 x 50 mm)
  • 4 x 150 mm initial™ brand green filled thermometers
  • 4 x 200 mm initial™ brand green filled thermometers
  • 2 x small classroom thermometers
  • 1 dial type maximum/minimum thermometer
  • 1 x soli temperature thermometer
  • 1 x Spotcheck clinical thermometer
Specifica
Include Blue plastic case, 4 x 150mm initial™ brand green filled thermometers, 4 x 200mm initial™ brand green filled thermometers, 2 x small classroom thermometers, 1 x dial type maximum/minimum thermometer, 1 x soli temperature thermometer, 1 x Spotcheck clinical thermometer
Altezza (sistema metrico) 50 mm
Larghezza (sistema metrico) 232 mm
Product Type Investigative Science Kit