Learn More
Thiamine-d3 Pyrophosphate Chloride (d3-Major), TRC
Specifications
Specifications
| Materiale o nome chimico | Thiamine-d3 Pyrophosphate Chloride |
| CAS | 2483831-79-2 |
| Note sul grado di purezza | HPLC |
| Formula molecolare | C12H16D3ClN4O7P2S |
| Quantità | 1 mg |
| Sinonimo | 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolium-d3 Chloride, Thiamine-d3 Trihydrogen Pyrophosphate Chloride, 3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolim-d3 Chloride P,P'-Dioxide, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)thiazolium-d3 Chloride, B-Neurox-d3, Berolase-d3, Biosyth-d3, Bioxilasi-d3, Bivitasi-d3, Co-B1-d3, Cocarbina-d3, Cocarboxil-d3, Cocarboxylase-d3, Coenbione-d3, Coxylase-d3, Diphosphothiamin-d3, Diphosphothiamine-d3, Farmaneurina-d3, Metabolase-d3, Nutrase-d3, Pyrolase-d3, Pyruvodehydrase-d3, TDP-d3, TDP (thiamin ester)-d3, TPP-d3, TPP (coenzyme), Thiamin-d3 Pyrophosphate, Thiamine-d3 Diphosphate, Thiamine-d3 Diphosphate Ester Chloride, Thiamine-d3 Diphosphoric Acid Ester Chloride, Thiamine-d3 Pyrophosphate, Thiamine-d3 Pyrophosphoric Acid Ester Chloride, Thiamine-d3 Pyrophosphoric Ester, Thiamine-PP-d3, Vitamin B1-d3 Pyrophosphate Chloride |
| InChI Formula | InChI=1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H/i2D3; |
| SMILES | OP(OP(OCCC1=C(C)[N+](CC2=CN=C(C([2H])([2H])[2H])N=C2N)=CS1)(O)=O)(O)=O.[Cl-] |
| Molecular Weight (g/mol) | 463.79 |
| Percent Purity | >95 |