Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Codice prodotto. 10689094
Cambia vista
Click to view available options
Dimensione del poro:
0.4 μm
1.0 μm
3 μm
3.0 μm
8.0 μm
No. of Wells:
6
12
24
Densità poro:
0.15 x 106 pores/cm2
0.6 x 106 pores/cm2
1 x 108 pores/cm2
2 x 106 pores/cm2
Dimensione della confezione:
pezzi 24
pezzi 48
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione del poro No. of Wells Densità poro unitSize
10689094 3 μm 6 0.6 x 106 pores/cm2 pezzi 24
10607184 0.4 μm 6 1 x 108 pores/cm2 pezzi 24
10053823 3 μm 24 2 x 106 pores/cm2 pezzi 48
10277463 0.4 μm 24 1 x 108 pores/cm2 pezzi 48
10093533 0.4 μm 12 2 x 106 pores/cm2 pezzi 48
10358623 0.4 μm 12 1 x 108 pores/cm2 pezzi 48
10044063 1.0 μm 12 2 x 106 pores/cm2 pezzi 48
10473845 3.0 μm 12 0.6 x 106 pores/cm2 pezzi 48
10742394 3.0 μm 12 2 x 106 pores/cm2 pezzi 48
10437573 8.0 μm 12 0.15 x 106 pores/cm2 pezzi 48
10443845 1.0 μm 24 2 x 106 pores/cm2 pezzi 48
10733544 3.0 μm 24 0.6 x 106 pores/cm2 pezzi 48
10679094 1.0 μm 6 2 x 106 pores/cm2 pezzi 24
10257033 3.0 μm 6 2 x 106 pores/cm2 pezzi 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Codice prodotto. 10689094 Fornitore Greiner Bio-One N. del fornitore 657630
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
TRUSTED_SUSTAINABILITY
Specifica
Materiale Polystyrene housing with sealed PET capillary pore membrane
Densità poro 0.6 x 106 pores/cm2
Dimensione del poro 3 μm
No. of Wells 6