Descrizione
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifica
Specifica
| Dimensione chiusura | S60/61 (m) to B83 (f) |
| Materiale | PTFE |
| Product Type | Thread Adapter |