Learn More
Idex Threaded Luer PEEK Adapter
Product Code. 12947759
Change view
Click to view available options
Color:
Black
Natural
For Use With (Equipment):
Female Luer x Female M6 Flat Bottom
Male Luer Lock x Female 10-32 Coned
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Unit Size
12947759 Black Female Luer x Female M6 Flat Bottom Each
12907759 Natural Male Luer Lock x Female 10-32 Coned Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12947759 Supplier Idex Supplier No. 0202359
  • ONLINE EXCLUSIVE PRICE
    34.00€
    31.17€ / pezzi 1
    Risparmia 2.83€
    8% Sconto
Spedizione stimata: 29-12-2026
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Type Threaded Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 125°C
Material PEEK
Color Black
For Use With (Equipment) Female Luer x Female M6 Flat Bottom