Learn More
Ticarcillin Disodium Salt (Contain 10% Methanol), TRC
Product Code. 30451838
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451838 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30451838 Supplier Toronto Research Chemicals Supplier No. TRCT4379011G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ticarcillin Disodium Salt
CAS 29457-07-6
Molecular Formula C15 H14 N2 O6 S2 . 2 Na
Quantity 1 g
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-2-carboxy-2-(3-thienyl)acetyl]amino]-3,3-dimethyl-7-oxo-','sodium salt (1:2)','(2S,5R,6R)-, 3-Thiophenemalonamic acid','N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-','disodium salt','D-(-)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(carboxy-3-thienylacetyl)amino]-3,3-dimethyl-7-oxo-','disodium salt','[2S-[2α,5α,6β(S*)]]-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-carboxy-3-thienylacetyl]amino]-3,3-dimethyl-7-oxo-','disodium salt','(2S,5R,6R)- (9CI), 6-D(-)-(α-Carboxy-3-thienylacetamido)penicillanic acid disodium salt, 6-D-(-)-α-Carboxy-3-thienylmethylacetamido penicillanic acid, AB 2288, BRL 2288, Disodium ticarcillin, Monapen, Ticar, Ticarcillin disodium, Ticarcillin disodium salt, Ticarcillin sodium, Ticarcillin sodium salt, Ticarpen, α-Carboxy-3-thienylmethylpenicillin disodium salt, D-Ticarcillin Disodium
InChI Formula InChI=1S/C15H16N2O6S2.2Na/c1-15(2)9(14(22)23)17-11(19)8(12(17)25-15)16-10(18)7(13(20)21)6-3-4-24-5-6;;/h3-5,7-9,12H,1-2H3,(H,16,18)(H,20,21)(H,22,23);;/q;2*+1/p-2/t7-,8-,9+,12-;;/m1../s1
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C(=O)[O-])c3ccsc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name disodium;(2S,5R,6R)-6-[[(2R)-2-carboxylato-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 428.39
Formula Weight 428.0089