missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, Alexa Fluor 488, Clone: 7D3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 567030
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometrySpecifica
TIM-3 (CD366) | |
Monoclonal | |
Alexa Fluor 488 | |
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
Human TIM-3 | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
7D3 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |