missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12008-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
TIMD4 is involved in encoding a phosphatidylserine receptor that enhances the engulfment of apoptotic cells. Involved in regulating T-cell proliferation and lymphotoxin signaling. Ligand for HAVCR1/TIMD1.Specifica
TIMD4 | |
Polyclonal | |
Unconjugated | |
TIMD4 | |
SMUCKLER, T cell membrane protein 4, TIM 4, TIM4, TIMD 4, TIMD4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91937 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96H15 | |
TIMD4 | |
TIMD4 Fusion Protein Ag2621 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |