Learn More
Timosaponin AIII, MedChemExpress
Product Code. 16473906
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473906 25 mg 25mg
16441518 5 mg 5mg
16409118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16473906 Supplier Medchem Express Supplier No. HYN0810
 Restricted Item
Spedizione stimata: 23-11-2026
MedChemExpress Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC50 of 35.4 μM.
Specifications
Chemical Name or Material Timosaponin AIII
CAS 41059-79-4
Color White
Purity Grade Notes Research
Molecular Formula C39H64O13
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (168.71 mM; Need ultrasonic)
SMILES C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@H](C)CO4)([H])C[C@@]1([H])[C@@](CC[C@@]5([H])[C@@]6(CC[C@H](O[C@@](O[C@H](CO)[C@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C5)C)([H])[C@]6([H])CC2
Molecular Weight (g/mol) 740.92
Formula Weight 740.92
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only