missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMP2 Polyclonal Antibody

Descrizione
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 79%.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Specifica
Specifica
| Antigene | TIMP2 |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.5 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TIMP2 |
| N. accesso geni | P25785 |
| Alias gene | Collagenase inhibitor; CSC-21K; D11Bwg1104e; DDC8; etID32613.12; fa96h09; Metalloproteinase inhibitor 2; metalloproteinase inhibitor 2a; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; metalloproteinase-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP metallopeptidase inhibitor 2a; TIMP2; Timp-2; TIMP-2 protein; timp2a; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase 2 precursor; tissue inhibitor of metalloproteinase 2a; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinase-2 precursor; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2; wu:fa96h09; zgc:136726 |
| Simboli geni | TIMP2 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?