Learn More
Lovibond™ Tintometer™ Alkalinity-Test Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca: Lovibond™ 515571BT
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
- Range: 20 to 800mg/L and 0.4 to 16mmol/L
- Parameter: Total alkalinity
- Method: Speed test method
- Tablet reagents have a minimum 5 year shelf life
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifica
Photometry Tablet Reagents | |
250 Tablets | |
Speed test method |
Test kits, colorimeters and spectrophotometers | |
20 To 800mg/L and 0.4 To 16mmol/L | |
Alkalinity-Test Tablets |