missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Bromocresol Purple Photometer Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
34.90€ - 76.30€
Specifica
Da utilizzare con (applicazione) | Photometry Tablet Reagents |
---|---|
Da utilizzare con (apparecchiatura) | Test kits, colorimeters and spectrophotometers |
Range | 5.2 to 6.8 |
Tipo | Bromocresol Purple Photometer Tablets |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15341875
|
Lovibond™
515700BT |
100 Tablets |
34.90€
pezzi 100 |
Spedizione stimata: 17-05-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
15351875
|
Lovibond™
515701BT |
250 Tablets |
76.30€
pezzi 250 |
Spedizione stimata: 17-05-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Range: 5.2 to 6.8
- Parameter: pH value
- Method: Bromcresol purple
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifica
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
5.2 to 6.8 | |
Bromocresol Purple Photometer Tablets |