Learn More
Lovibond™ Tintometer™ DPD No. 3 Nessler Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca:  Lovibond™ 511250BT
Dettagli aggiuntivi : Peso : 0.30000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15331945
  • 38.30€ / pezzi 100
Spedizione stimata
29-06-2020
Aggiungi al carrello
Descrizione
Descrizione
  • Available in Packs of 100 and 2550 Tablets
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifica
Specifica
100 Tablets
Test kits, colorimeters and spectrophotometers
Photometry Tablet Reagents
DPD No. 3 Nessler Tablets
Water Testing