Learn More
Lovibond™ Tintometer™ Fluoride Excess Al Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca: Lovibond™ 511410
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Fluoride Excess Al Tablets
- Available in packs of 100 and 250
Specifica
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Fluoride Excess Al Tablets |