Learn More
Lovibond™ Tintometer™ Molybdate No. 2 HR Tablet
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca:  Lovibond™ 513071BT
Dettagli aggiuntivi : Peso : 0.30000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15321785
  • 99.40€ / pezzi 250
Spedizione stimata: 14-05-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Range: 1 to 50mg/L
  • Parameter: Molybdate LR, molybdate, HR molybdate
  • Method: Thioglycolate
  • For use in our test kits, colorimeters and spectrophotometers
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifica
Specifica
Photometry Tablet Reagents
250 Tablets
Molybdate No. 2 HR Tablet
Test kits, colorimeters and spectrophotometers
1 to 50mg/L