Learn More
Lovibond™ Tintometer™ Organo Phosphonate No. 1 Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
40.00€ - 88.30€
Specifica
Da utilizzare con (applicazione) | Photometry Tablet Reagents |
---|---|
Da utilizzare con (apparecchiatura) | Test kits, colorimeters and spectrophotometers |
Tipo | Organo Phosphonate No. 1 Tablets |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15321775
|
Lovibond™
512961BT |
250 Tablets |
88.30€
pezzi 250 |
Spedizione stimata: 17-05-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
15311775
|
Lovibond™
512960BT |
100 Tablets |
Questo articolo non è attualmente disponibile o è stato dismesso. La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative. |
||||||
Descrizione
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifica
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
Organo Phosphonate No. 1 Tablets |