Learn More
Lovibond™ Tintometer™ Thymol Blue Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca:  Lovibond™ 511650BT
Dettagli aggiuntivi : Peso : 0.30000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15321735
  • 34.90€ / pezzi 100
Spedizione stimata: 26-04-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Thymol Blue Tablets
  • Available in packs of 100 and 250
Specifica
Specifica
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
100 Tablets
Thymol Blue Tablets