Descrizione
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da
Specifica
Specifica
| Product Line | Novus |
| Colore | White |
| Content And Storage | TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da |
| Da utilizzare con (applicazione) | Inhibit TLR2 and TLR4 signaling |
| Product Type | TIRAP (TLR2 and TLR4) Inhibitor Peptide Set |
| Quantità | 1 mg |
For Research Use Only