Learn More
Macherey-Nagel™ TLC Simultaneous Developing Chamber
Product Code. 10486471 Shop All Macherey-Nagel Products
Change view
Click to view available options
Holds:
2
5
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10486471 5 Each
12374613 2 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10486471 Supplier Macherey-Nagel™ Supplier No. 814019
  • ONLINE EXCLUSIVE PRICE
    464.00€
    431.55€ / pezzi 1
    Risparmia 32.45€
    6% Sconto
View all promotions
In stock
Add to basket
Edge
Add to basket
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
  • Accessory for TLC
  • Chambers are thick walled
  • Clear glass tank 
  • Heavy ground glass lid
Specifications
Quantity 1 Pk.
Type TLC Simultaneous Developing Chamber
For Use With (Application) For 5 Plates 20 x 20 cm
Holds 5