missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 (Synovial Sarcoma Marker) Mouse, Clone: TLE1/2051, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 7088-MSM1-P0
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TLE1 (Synovial Sarcoma Marker) | |
Nuclear | |
TLE1/2051 | |
Unconjugated | |
Mouse | |
83kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7088 | |
IgG2a κ |
Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human TLE1 fragment (aa 175-338) | |
HeLa, HepG2 or Jurkat cells. Endometrium Carcinoma or Sarcomas | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |