missing translation for 'onlineSavingsMsg'
Learn More

TMED5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA562164

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15759961

  • 512.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: FELILDNMGE QAQEQEDWKK YITGTDILDM KLEDILESIN SIKSRLSKSG HIQTLL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

TMED5 Polyclonal antibody specifically detects TMED5 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifica

Specifica

TMED5
Polyclonal
Unconjugated
TMED5
CGI-100; p24 family protein gamma-2; p24g2; p24gamma2; p28; TMED5; Transmembrane emp24 domain-containing protein 5; transmembrane emp24 protein transport domain containing 5; transmembrane p24 trafficking protein 5; UNQ397/PRO733
Rabbit
Antigen affinity chromatography
RUO
50999
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y3A6
TMED5
Recombinant protein corresponding to Human TMED5
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only