Learn More
TMEM38A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562194
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: THSHSSPFDA LEGYICPVLF GSACGGDHHH DNHGGSHSGG GPGAQHSAMP AKSKEELSEG SRKK Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
TMEM38A Polyclonal antibody specifically detects TMEM38A in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
TMEM38A | |
Polyclonal | |
Unconjugated | |
Tmem38a | |
1110001E17Rik; 27 kDa sarcoplasmic reticulum protein; AI413399; mg33a; Mitsugumin33A; Mitsugumin-33A; RGD1307901; sarcoplasmic reticulum protein 27; SPR-27; Srp-27; TMEM38A; tmem38a {ECO:0000250; Transmembrane protein 38A; TRICA; TRIC-A; trimeric intracellular cation channel type A; UniProtKB:Q3TMP8} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79041 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6F2 | |
Tmem38a | |
Recombinant protein corresponding to Human TMEM38A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only