Learn More
SUCNR1/GPR91 Antibody - BSA Free, Novus Biologicals™
Codice prodotto. 18740294 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.025 mg
0.1 mg
Dimensione della confezione:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18740294 0.1 mg 0.10mg
18787863 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18740294 Fornitore Novus Biologicals N. del fornitore NBP100861
Rabbit Polyclonal Antibody has been used in 14 publications
SUCNR1/GPR91 Polyclonal specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay.
TRUSTED_SUSTAINABILITY
Specifica
Antigene SUCNR1/GPR91
Applicazioni Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Concentrazione 1.0 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 1-2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, In vitro assay reported in scientific literature (PMID 30478422)
Formulazione PBS with 0.02% Sodium Azide
N. accesso geni Q9BXA5
Alias gene GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1
Simboli geni SUCNR1
Specie ospite Rabbit
Immunogeno A synthetic peptide made to an internal portion of the human GPR91 protein (between residues 100-200) [Uniprot: Q9BXA5]
Peso molecolare dell'antigene 38 kDa
Metodo di purificazione Affinity Purified
Quantità 0.1 mg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 56670
Test di specificità GPR91 (A135) detects endogenous levels of GPR91 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
For Research Use Only