Learn More
α-Tocopherol phosphate, MedChemExpress
Product Code. 16459667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459667 5 mg 5mg
16457128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16459667 Supplier Medchem Express Supplier No. HY16686
 Restricted Item
Spedizione stimata: 15-12-2026
MedChemExpress α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.
α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.
Specifications
Chemical Name or Material α-Tocopherol phosphate
CAS 38976-17-9
Color White
Purity Grade Notes Research
Molecular Formula C29H51O5P
Quantity 5 mg
Synonym alpha-Tocopherol phosphate TocP vitamin E phosphate
Solubility Information DMSO : ≥ 100 mg/mL (195.81 mM) ∣H2O : 0.67 mg/mL (1.31 mM; Need ultrasonic)
SMILES CC1=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C(C)C(OP(O)(O)=O)=C1C
Molecular Weight (g/mol) 510.69
Formula Weight 510.69
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only