Learn More
Tocris Bioscience™ Psalmotoxin 1
Codice prodotto. 18739267
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18739267 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18739267 Fornitore Tocris Bioscience™ N. del fornitore 5042/100U
Potent and selective ASIC1a channel blocker
Psalmotoxin 1 is a potent and selective acid-sensing ion channel 1a (ASIC1a) blocker (IC50 = 0.9 nM). Displays no effect at ASIC1b, ASIC2a, ASIC3, heteromeric ASIC channels, ENaC and KV2.1/2.2/4.2/4.3 channels expressed in oocytes, at concentrations up to 100 nM. Displays potent analgesic properties against thermal, mechanical, chemical, inflammatory and neuropathic pain in rodents.
TRUSTED_SUSTAINABILITY
Specifica
CAS 316808-68-1
Quantità 100 μg