missing translation for 'onlineSavingsMsg'
Learn More

TOMM22 Recombinant Protein, Abnova

Marca:  Abnova H00056993-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16162403

  • 458.00€ / 25µg


Spedizione stimata
19-11-2020
Aggiungi al carrello

Descrizione

Descrizione

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH09363
Solution
56993
TOMM22 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TOMM22
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
41.36
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAAAVAAAGAGEPQSPDELLPKGDAEKPEEELEEDDDEELDETLSERLWGLTEMFPERVRSAAGATFDLSLFVAQKMYRFSRAALWIGTTSFMILVLPVVFETEKLQMEQQQQLQQRQILLGPNTGLSGGMPGALPSLPGKI
1C9-2|MST065|MSTP065|TOM22
TOMM22
Wheat Germ (in vitro)
GST
Videos
Documenti