Learn More
Tonalide, TRC
Product Code. 30457228
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457228 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457228 Supplier Toronto Research Chemicals Supplier No. TRCT53980050MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tonalide
CAS 21145-77-7
Molecular Formula C18 H26 O
Quantity 50 mg
Synonym 2'-Acetonaphthone','5',6',7',8'-tetrahydro-3',5',5',6',8',8'-hexamethyl- (8CI), 1-(5,6,7,8-Tetrahydro-3,5,5,6,8,8-hexamethyl-2-naphthalenyl)ethanone, 6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, 6-Acetyl-1,1,2,4,4,7-hexamethyltetraline, 7-Acetyl-1,1,3,4,4,6-hexamethyltetralin, 7-Acetyl-1,1,3,4,4,6-hexamethyltetraline, AHMT, AHMT (perfume), AHTN, Acetylhexamethyltetrahydronaphthalene, Extralide, Fixolide, Musk tonalid, NSC 19550, Tentarome, Tetralide, Tonalid, Tonalide
InChI Formula InChI=1S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12H,10H2,1-7H3
SMILES CC1CC(C)(C)c2cc(C(=O)C)c(C)cc2C1(C)C
IUPAC Name 1-(3,5,5,6,8,8-hexamethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol) 258.4
Formula Weight 258.1984