missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOPBP1 Monoclonal Antibody (6D12), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524436
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Peptide Sequence: LLQSGGAKVL PGHSVPLFKE ATHLFSDLNK LKPDDSGVNI AEAAAQNVYC LRTEYIADYL MQESPPHVEN YCLPEAISFI QNNKELGTGL SQKRKAPTEK NKIKRPRVH
TOPBP1 Monoclonal antibody specifically detects TOPBP1 in Human samples. It is validated for ELISA, Western Blot.Specifica
TOPBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q92547 | |
TOPBP1 | |
TOPBP1 (NP_008958, 1327 a.a. ∽ 1435 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
6D12 | |
Unconjugated | |
TOPBP1 | |
1110031N14Rik; 2810429C13Rik; AI256758; D430026L04Rik; DNA topoisomerase 2-binding protein 1; DNA topoisomerase II-beta-binding protein 1; DNA topoisomerase II-binding protein 1; KIAA0259; mKIAA0259; RGD1562949; TOP2BP1; TOPBP 1; Topbp1; topoisomerase (DNA) II beta binding protein; topoisomerase (DNA) II binding protein 1 | |
Mouse | |
Protein A | |
RUO | |
11073 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |