Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Data Logging Thermohygrometers with Calibration
Codice prodotto. 17367649 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Dimensioni (L x P x A):
14 x 2.5 x 8.3 cm
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P x A) Dimensione della confezione
17367649 14 x 2.5 x 8.3 cm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17367649 Fornitore Fisherbrand™ N. del fornitore 9876716
  • ONLINE EXCLUSIVE PRICE
    349.00€
    282.54€ / pezzi 1
    Risparmia 66.46€
    19% Sconto
Spedizione stimata: 30-12-2026
The Traceable™ Memory-Loc™ Data Logging Thermohygrometer with Calibration allows you to safely secure collected information — data cannot be overwritten or modified.
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory.
  • Store up to 30 unique alarm events.
  • Smart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditions.
  • Adjustable sampling rate — from 1 min. to 12 hr. in 1 min. steps.
  • Can upload a USB downloaded file and store on a TraceableLIVE™ cloud account.
Specifica
Tipo Thermohygrometer
Intervallo di umidità 0 to 95%
Risoluzione 0.1
Temperature Range (Metric) -20°C to +70°C
Precisione termica ±0.4°C
Descrizione Traceable™ Memory-Loc™ Thermohygrometer with Calibration
Memoria 688,086 Readings
Dimensioni (L x P x A) 14 x 2.5 x 8.3 cm