missing translation for 'onlineSavingsMsg'
Learn More

trans-1-n-Pentyl-4-(4-n-propylcyclohexyl)benzene, 99%, Thermo Scientific™

Catalog Number 15442696
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
89363-50-8
C20H32
272.476
MFCD22125193
RSUADYMBPNVXJC-UHFFFAOYSA-N
1-pentyl-4-trans-4-propylcyclohexyl benzene, 1-4-propylcyclohexyl-4-pentylbenzene, 4-trans-4-propylcyclohexyl pentylbenzene, 4-trans-4-propylcyclohexyl-pentylbenzene, 1-pentyl-4-4-propylcyclohexyl benzene, trans-1-n-pentyl-4-4-n-propylcyclohexyl benzene, 1-pentyl-4-1s,4r-4-propylcyclohexyl benzene
15606764
1-pentyl-4-(4-propylcyclohexyl)benzene
CCCCCC1=CC=C(C=C1)C2CCC(CC2)CCC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
89363-50-8
C20H32
272.476
MFCD22125193
RSUADYMBPNVXJC-UHFFFAOYSA-N
1-pentyl-4-trans-4-propylcyclohexyl benzene, 1-4-propylcyclohexyl-4-pentylbenzene, 4-trans-4-propylcyclohexyl pentylbenzene, 4-trans-4-propylcyclohexyl-pentylbenzene, 1-pentyl-4-4-propylcyclohexyl benzene, trans-1-n-pentyl-4-4-n-propylcyclohexyl benzene, 1-pentyl-4-1s,4r-4-propylcyclohexyl benzene
15606764
1-pentyl-4-(4-propylcyclohexyl)benzene
CCCCCC1=CC=C(C=C1)C2CCC(CC2)CCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico trans-1-n-Pentyl-4-(4-n-propylcyclohexyl)benzene
Quantità 1 g
Peso formulazione 272.47
Percent Purity 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'