Learn More
trans-3'-Hydroxy Cotinine N-Beta-D-Glucuronide, TRC
Product Code. 30138139
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138139 5 mg 5mg
30132850 2 mg 2mg
30008932 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138139 Supplier Toronto Research Chemicals Supplier No. TRCH9245305MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 146275-18-5
Molecular Formula C16 H20 N2 O8
Molecular Weight (g/mol) 368.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]-, inner salt (9CI, ACI), Pyridinium, 1-β-D-glucopyranuronosyl-3-(4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl)-, inner salt, (2S-trans)- (ZCI), trans-3'-Hydroxycotinine-N-glucuronide, trans-3'-Hydroxy Cotinine N-β-D-Glucuronide, 1-β-D-Glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]pyridinium inner salt, (3R,5S)-3-Hydroxy-1-methyl-5-(3'-pyridinyl)-2-pyrrolidinone N'-β-D-Glucuronide, trans-3'-Hydroxycotinine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[(3R,5S)-1-methyl-2-oxo-5-pyridin-3-ylpyrrolidin-3-yl]oxyoxane-2-carboxylic acid
SMILES CN1[C@@H](C[C@@H](O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C1=O)c3cccnc3
Tekniske data
Quantity 5 mg
InChI Formula InChI=1S/C16H20N2O8/c1-18-8(7-3-2-4-17-6-7)5-9(14(18)22)25-16-12(21)10(19)11(20)13(26-16)15(23)24/h2-4,6,8-13,16,19-21H,5H2,1H3,(H,23,24)/t8-,9+,10-,11-,12+,13-,16+/m0/s1
Formula Weight 368.122
Chemical Name or Material trans-3'-Hydroxycotinine-N-beta-D-glucuronide
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.