missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-7 siRNA
Blocks transcription and translation of Caspase-7 from Human.
Marca: enQuireBio™ QX42-2nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001218 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
840 | |
2 nmol | |
Human |
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. | |
CASP7 | |
siRNA | |
ICE-LAP3, CMH-1, CASP-7, MCH3, LICE2, CASP7 | |
Lyophilized |