missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human IRF1 siRNA
Blocks transcription and translation of IRF1 from Human.
Marca: enQuireBio™ QX33-2nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002189 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3659 | |
2 nmol | |
Human |
U-2 OS Huan Osteosarcoma Epithelial Cells from Bone. | |
IRF1 | |
siRNA | |
IRF-1, Interferon Regulatory Factor 1, MAR | |
Lyophilized |