Learn More
Uvex Trend BOA™ Black/Gray/Red PU/PU 69083 Low Shoe - Width 12
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Marca:  Uvex 6908338
Dettagli aggiuntivi : Peso : 1.04800kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17253311
  • 107.00€ / 1 paio
Spedizione stimata: 27-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 6908.3 S1 P SRC
  • Sporty, S1 P perforated safety shoe with wide fit
  • Breathable perforated microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
    • Helps maintain foot temperature for optimum wearer comfort
  • Soft padding on collar and tongue
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 6908.3
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
Specifica
Specifica
Red, Gray, Black
38
Steel
EN ISO 20345:2011
Wide width
Yes
S1
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC