missing translation for 'onlineSavingsMsg'
Learn More

TRF-1 Antibody (3H11), mFluor Violet 610 SE, Novus Biologicals™

Codice prodotto. 18743143 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18743143 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18743143 Fornitore Novus Biologicals N. del fornitore NB12014397MFV610

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

TRF-1 Monoclonal antibody specifically detects TRF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifica

Antigene TRF-1
Applicazioni Western Blot, Immunohistochemistry, Immunocytochemistry
Classificazione Monoclonal
Clone 3H11
Coniugato mFluor Violet 610 SE
Diluizione Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulazione 50mM Sodium Borate
Alias gene NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Specie ospite Mouse
Immunogeno Human TRF-1
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 7013
Target Species Human
Content And Storage Store at 4C in the dark.
Forma Purified
Isotype IgG1 κ
Vedi altri risultati Mostra meno risultati
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.