Learn More
Trichloroacetic acid, Chem Lab™
Product Code. 11964064
Change view
Click to view available options
Quantity:
200 g
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11964064 200 g 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11964064 Supplier Chem-Lab Supplier No. CL02.2003.1000
  • ONLINE EXCLUSIVE PRICE
    132.00€
    69.90€ / 1L
    Risparmia 62.10€
    47% Sconto
 Restricted Item
Spedizione stimata: 28-12-2026
Specifications
Concentration 20%
Density 1.12g/mL
Quantity 200 g
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid 20% w/v solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Trichloroacetic acid