Learn More
Trichloromethyl Chloroformate, TRC
Specifica
Specifica
| Materiale o nome chimico | Trichloromethyl Chloroformate |
| CAS | 503-38-8 |
| Formula molecolare | C2Cl4O2 |
| Quantità | 10 g |
| Sinonimo | Carbonochloridic Acid Trichloromethyl Ester, Chloroformic Acid Trichloromethyl Ester, Trichloromethanol Chloroformate, Diphosgen, Diphosgene, Trichloromethyl Chlorocarbonate |
| InChI Formula | InChI=1S/C2Cl4O2/c3-1(7)8-2(4,5)6 |
| SMILES | ClC(=O)OC(Cl)(Cl)Cl |
| IUPAC Name | trichloromethyl carbonochloridate |
| Molecular Weight (g/mol) | 197.83 |
| Peso formulazione | 195.8652 |