Learn More
Triethylamine Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Triethylamine Hydrochloride |
| CAS | 554-68-7 |
| Formula molecolare | C6H16ClN |
| Quantità | 25 g |
| Sinonimo | Triethylamine','Hydrochloride, N,N-Diethylethanamine Hydrochloride, Triethylamine Monohydrochloride, Triethylammonium Chloride |
| InChI Formula | InChI=1S/C6H15N.ClH/c1-4-7(5-2)6-3;/h4-6H2,1-3H3;1H |
| SMILES | Cl.CCN(CC)CC |
| IUPAC Name | N,N-diethylethanamine;hydrochloride |
| Molecular Weight (g/mol) | 137.65 |
| Peso formulazione | 137.0971 |