Descrizione
- Finest quality products for life science
- The most stringent testing protocols and highest purity
- Superior manufacturing methods and product specific testing protocols designed to ensure quality to our highest standard
- Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
Specifica
Specifica
| CAS | 10138-93-9 |
| pH | 2.9 to 3 |
| Quantità | 100 mL |
| Imballaggio | Glass bottle |
| Molecular Weight (g/mol) | 199.187 |