Learn More
Trimethyl borate, 99%, Thermo Scientific Chemicals
Catalog Number p-7024943
Click to view available options
Quantità:
250 mL
1000 mL
121-43-7
C3H9BO3
103.912
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
121-43-7
C3H9BO3
103.912
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.
Solubility
Miscible with tetrahydrofuran, ether, isoporpylamine, hexane and methanol.
Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It decomposes slowly in the presence of moisture and rapidly in water.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Trimethyl borate
Punto di fusione -34°C
Densità 0.915
Punti di ebollizione 67°C to 69°C
Punto d'infiammabilità −8°C (17°F)
Odore Odorless
Formula lineare (CH3O)3B
Indice di rifrazione 1.361
Quantità 250 mL
Numero UN UN2416
Beilstein 1697939
Sensibilità Moisture sensitive
Indice di Merck 14,9712
Informazioni di solubilità Miscible with tetrahydrofuran,ether,isoporpylamine,hexane and methanol.
Peso formulazione 103.92
Percent Purity 99%