Learn More
Trimethyl(bromodifluoromethyl)silane, TRC
Specifica
Specifica
| Materiale o nome chimico | Trimethyl(bromodifluoromethyl)silane |
| CAS | 115262-01-6 |
| Formula molecolare | C4H9F2SiBr |
| Quantità | 1 g |
| Sinonimo | (Bromodifluoromethyl)trimethylsilane |
| InChI Formula | InChI=1S/C4H9BrF2Si/c1-8(2,3)4(5,6)7/h1-3H3 |
| SMILES | C[Si](C)(C)C(Br)(F)F |
| Molecular Weight (g/mol) | 203.1 |