Learn More
Trimethyl Phosphate-d9, TRC
Specifica
Specifica
| Materiale o nome chimico | Trimethyl Phosphate-d9 |
| CAS | 32176-12-8 |
| Formula molecolare | C3 D9 O4 P |
| Quantità | 250 mg |
| Sinonimo | Trimethyl phosphate D9, Phosphoric acid','tri(methyl-d3) ester (8CI), Methan-d3-ol','phosphate (3:1) (9CI), NSC 58985-d9, TMP-d9, TMPA-d9, TMPO-d9, Trimethoxyphosphine Oxide-d9, Trimethyl Orthophosphate-d9, Trimethyl Phosphate-d9 |
| InChI Formula | InChI=1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3/i1D3,2D3,3D3 |
| SMILES | [2H]C([2H])([2H])OP(=O)(OC([2H])([2H])[2H])OC([2H])([2H])[2H] |
| IUPAC Name | tris(trideuteriomethyl) phosphate |
| Molecular Weight (g/mol) | 149.13 |
| Peso formulazione | 149.0803 |