Learn More
Trimethylphenylammonium-d9 Iodide, TRC
Specifica
Specifica
| Materiale o nome chimico | Trimethylphenylammonium-d9 Iodide |
| CAS | 88278-22-2 |
| Formula molecolare | C9H5D9IN |
| Quantità | 100 mg |
| Sinonimo | Trimethylanilinium-d9 Iodide, Phenyltrimethylammonium-d9 Iodide, N,N,N-Trimethylanilinium-d9 Iodide |
| InChI Formula | InChI=1S/C9H14N.HI/c1-10(2,3)9-7-5-4-6-8-9;/h4-8H,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3; |
| SMILES | [I-].[2H]C([2H])([2H])[N+](c1ccccc1)(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | phenyl-tris(trideuteriomethyl)azanium;iodide |
| Molecular Weight (g/mol) | 272.17 |
| Peso formulazione | 272.0736 |