Learn More
Trimethylsulfonium Chloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Trimethylsulfonium Chloride |
| CAS | 3086-29-1 |
| Formula molecolare | C3H9S . Cl |
| Quantità | 500 mg |
| Sinonimo | Sulfonium','trimethyl-','chloride (1:1) (ACI), Sulfonium','trimethyl-','chloride (8CI','9CI), Trimethylsulfonium chloride (6CI','7CI) |
| InChI Formula | InChI=1S/C3H9S.ClH/c1-4(2)3;/h1-3H3;1H/q+1;/p-1 |
| SMILES | [Cl-].C[S+](C)C |
| IUPAC Name | trimethylsulfanium;chloride |
| Molecular Weight (g/mol) | 112.62 |
| Peso formulazione | 112.0113 |