missing translation for 'onlineSavingsMsg'
Learn More

TRIS-HEPES-native running buffer (10X), pH 8.0, Thermo Scientific™

Catalog Number p-7076594
Click to view available options
Quantità:
500 mL
1 L
2 L
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico TRIS-HEPES-native running buffer (10X)
Imballaggio Plastic bottle
Quantità 500 mL
Forma fisica Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'