missing translation for 'onlineSavingsMsg'
Learn More

Tropine DL-tropate, 99%, Thermo Scientific Chemicals

Catalog Number p-7082418
206.67 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51-55-8
C17H23NO3, C17H23NO3
DL-Hyoscyamine; Atropine
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51-55-8
C17H23NO3, C17H23NO3
DL-Hyoscyamine; Atropine

Reduces brain levels of acetylcholine

Tropine DL-tropate is used as a reduces brain levels of acetylcholine, Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors, used in opthalmology to dilate the pupil and in efforts of resuscitation in the treatment of bradycardia and cardiac arrest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantità 25 g
Descrizione White
Da utilizzare con (applicazione) Reduces brain levels of acetylcholine
Saggio 99%
Peso formulazione 289.37g/mol
Sensibilità Light Sensitive
Product Type Signal Transduction Reagent
Forma Powder
Beilstein 91260
InChI Key RKUNBYITZUJHSG-UHFFFAOYNA-N
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate
Molecular Weight (g/mol) 289.38
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'