Learn More
Trospium Chloride, Mikromol™
Product Code. 17920376
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17920376 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17920376 Supplier Mikromol Supplier No. MM1266.00
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trospium Chloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trospium chloride
CAS 10405-02-4
Analyte or Component Names Trospium chloride
Shipping Condition Room Temperature
Molecular Formula C25 H30 N O3 . Cl
Certifications/Compliance ISO 17034
Synonym Trospium chloride, (1R,3r,5S)-3-[(Hydroxydiphenylacetyl)oxy]spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride, As XVII, Azoniaspiro compound XVII, Keptan, Relaspium, Sanctura, Sanctura XR, Spasmex, Spasmo 3, Spasmo-lyt, Trozyd, Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-, chloride (1:1), (1alpha,3beta,5alpha)-, Spiro[1alphaH,5alphaH-nortropane-8,1'-pyrrolidinium], 3alpha-hydroxy-, chloride, benzilate (8CI), Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], 3-[(hydroxydiphenylacetyl)oxy]-, chloride, (1alpha,3beta,5alpha)- (9CI), Benzilic acid, ester with 3alpha-hydroxyspiro[1alphaH,5alphaH-nortropane-8,1'-pyrrolidinium] chloride (8CI), Azoniaspiro(3alpha-benziloyloxynortropane-8,1'-pyrrolidine) chloride
InChI Formula InChI=1S/C25H30NO3.ClH/c27-24(25(28,19-9-3-1-4-10-19)20-11-5-2-6-12-20)29-23-17-21-13-14-22(18-23)26(21)15-7-8-16-26;/h1-6,9-12,21-23,28H,7-8,13-18H2;1H/q+1;/p-1/t21-,22+,23+;
SMILES [Cl-].OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)[N+]23CCCC3)(c4ccccc4)c5ccccc5
IUPAC Name [(1S,5R)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
Molecular Weight (g/mol) 427.96
Quantity 250 mg
Formula Weight 427.1914 g/mol